4-(Hexyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
H1357-5G
H1357-1G
TCH1357-5GEA
213.3
CAD
TCH1357-5G
TCH1357-1G
4-(Hexyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Hexyloxy)phenylboronic acid
CAS Number: 121219-08-7
MDL Number: MFCD04039027
Molecular Formula: C12H19BO3
Molecular Weight: 222.09
Form: Crystal
Color: White
Melting point (°C): 85
Other Product Names / Synonyms: 4-Hexyloxybenzeneboronic Acid
Formula:
C₁₂H₁₉BO₃ Melting Pt: 85 °C |
MDL Number:
MFCD04039027 CAS Number: 121219-08-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...