Hexaphenoxycyclotriphosphazene ≥98.0%

Supplier: TCI America

Synonyms: Phenoxycycloposphazene

H1356-25G H1356-5G
TCH1356-25GEA 1106 CAD
TCH1356-25G TCH1356-5G
Hexaphenoxycyclotriphosphazene ≥98.0%
Hexaphenoxycyclotriphosphazene

CAS Number: 1184-10-7
MDL Number: MFCD00183774
Molecular Formula: C36H30N3O6P3
Molecular Weight: 693.57
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 114
Lambda max.: 261 nm (CHCl3)

Formula: C₃₆H₃₀N₃O₆P₃
MW: 693.57 g/mol
Boiling Pt: 280 °C
Melting Pt: 114 °C
MDL Number: MFCD00183774
CAS Number: 1184-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bce66e91c53fadacc4d0edf366f234135e246e6c&vid=bce66e91c53fadacc4d0edf366f234135e246e6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR