7-Hydroxycoumarin-3-carboxylic acid ≥98.0% (by HPLC)

Supplier: TCI America

H1352-1G H1352-200MG
TCH1352-1GEA 974.86 CAD
TCH1352-1G TCH1352-200MG
7-Hydroxycoumarin-3-carboxylic acid ≥98.0% (by HPLC)
7-Hydroxycoumarin-3-carboxylic acid

CAS Number: 779-27-1
MDL Number: MFCD00017491
Molecular Formula: C10H6O5
Molecular Weight: 206.15
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 263
Storage Temperature: <0°C


Product Categories: Bioscience, Donors & Acceptors for Fluorescent Resonance Energy Transfer (FRET), Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Umbelliferone-3-carboxylic Acid

Formula: C₁₀H₆O₅
Melting Pt: 263 °C
MDL Number: MFCD00017491
CAS Number: 779-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6812a3197138aecf3bd97e75b9513a827969811b&vid=6812a3197138aecf3bd97e75b9513a827969811b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR