2-Hexylthiophene ≥98.0% (by GC)
Supplier: TCI America
H1350-25G
H1350-5G
TCH1350-25GEA
331.8
CAD
TCH1350-25G
TCH1350-5G
2-Hexylthiophene ≥98.0% (by GC)
2-Hexylthiophene
CAS Number: 18794-77-9
MDL Number: MFCD00022535
Molecular Formula: C10H16S
Molecular Weight: 168.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 100
Specific Gravity (20/20): 0.94
Lambda max.: 234 nm (EtOH)
Formula:
C₁₀H₁₆S Boiling Pt: 92 °C Density: 0.94 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00022535 CAS Number: 18794-77-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...