3-Hydroxy-5-methylisoxazole ≥98.0% (by GC)

Supplier: TCI America

H1348-25G H1348-5G
TCH1348-25GEA 218.04 CAD
TCH1348-25G TCH1348-5G
3-Hydroxy-5-methylisoxazole ≥98.0% (by GC)
3-Hydroxy-5-methylisoxazole

CAS Number: 10004-44-1
MDL Number: MFCD00144468
Molecular Formula: C4H5NO2
Molecular Weight: 99.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86
Storage Temperature: <0°C


Product Categories: Bioscience, Pesticide Ingredients, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Hymexazol

Formula: C₄H₅NO₂
Melting Pt: 86 °C
MDL Number: MFCD00144468
CAS Number: 10004-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0711943a821418de622e8c967681dce494ff0925&vid=0711943a821418de622e8c967681dce494ff0925&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR