4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl ≥97.0% (by titrimetric analysis)

Supplier: TCI America

H1347-1G H1347-200MG
TCH1347-1GEA 841.35 CAD
TCH1347-1G TCH1347-200MG
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl ≥97.0% (by titrimetric analysis)
4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl

CAS Number: 81998-04-1
MDL Number: MFCD02916382
Molecular Formula: C12H12N2O
Molecular Weight: 200.24
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 121


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4-Hydroxymethyl-4'-methyl-2,2'-bipyridine, 4'-Methyl-2,2'-bipyridine-4-methanol

Formula: C₁₂H₁₂N₂O
Melting Pt: 121 °C
MDL Number: MFCD02916382
CAS Number: 81998-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3cbcaf06c5b2863aa7097ebe15a287b632f327d&vid=f3cbcaf06c5b2863aa7097ebe15a287b632f327d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR