Homophthalic anhydride ≥98.0% (by GC)
Supplier: TCI America
H1346-25G
H1346-5G
TCH1346-25GEA
357.08
CAD
TCH1346-25G
TCH1346-5G
Homophthalic anhydride ≥98.0% (by GC)
Homophthalic anhydride
CAS Number: 703-59-3
MDL Number: MFCD00006894
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 142
Other Product Names / Synonyms: 1,3-Isochromandione
Formula:
C₉H₆O₃ Boiling Pt: 161 °C Melting Pt: 142 °C |
MDL Number:
MFCD00006894 CAS Number: 703-59-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...