2-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

H1345-1G H1345-5G
TCH1345-1GEA 56.25 CAD
TCH1345-1G TCH1345-5G
2-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Hydroxymethyl)phenylboronic acid

CAS Number: 87199-14-2
MDL Number: MFCD02258966
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-(Hydroxymethyl)benzeneboronic Acid

Formula: C₇H₉BO₃
MDL Number: MFCD02258966
CAS Number: 87199-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0696a13a9740cd7d566bc4dfc730d8d991bd2fc0&vid=0696a13a9740cd7d566bc4dfc730d8d991bd2fc0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR