Salicylic acid ≥99.5% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

H1342-25G H1342-5G
TCH1342-25GEA 75.84 CAD
TCH1342-25G TCH1342-5G
Salicylic acid ≥99.5% (by titrimetric analysis) for biochemical research
Salicylic acid

[for Biochemical Research]
CAS Number: 69-72-7
MDL Number: MFCD00002439
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >99.5% (T)
Form: Crystal
Boiling point (°C): 211
Melting point (°C): 159
Flash Point (°C): 157


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Salicylic Acid

Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211 °C
Melting Pt: 159 °C
Flash Pt: 157 °C
MDL Number: MFCD00002439
CAS Number: 69-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9353f7949263aec8589db1452164f4ad85771fb1&vid=07bd493cb2bade9dbe727bb6e940f6876a28c9ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR