(R)-2-Hydroxy-4-phenylbutyric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H1338-1G
H1338-5G
TCH1338-1GEA
170.64
CAD
TCH1338-1G
TCH1338-5G
(R)-2-Hydroxy-4-phenylbutyric acid ≥98.0% (by titrimetric analysis)
(R)-2-Hydroxy-4-phenylbutyric acid
CAS Number: 29678-81-7
MDL Number: MFCD00192219
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 117
Specific rotation [a]20/D: -8 deg (C=1, EtOH)
Formula:
C₁₀H₁₂O₃ Melting Pt: 117 °C |
MDL Number:
MFCD00192219 CAS Number: 29678-81-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...