trans-10-Hydroxy-2-decenoic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (E)-10-Hydroxydec-2-enoic acid, Royal Jelly acid

H1337-1G H1337-200MG
TCH1337-1GEA 379.2 CAD
TCH1337-1G TCH1337-200MG
trans-10-Hydroxy-2-decenoic acid ≥97.0% (by titrimetric analysis)
trans-10-Hydroxy-2-decenoic acid

CAS Number: 14113-05-4
MDL Number: MFCD00204506
Molecular Formula: C10H18O3
Molecular Weight: 186.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 66
Storage Temperature: 0-10°C

Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
Melting Pt: 66 °C
MDL Number: MFCD00204506
CAS Number: 14113-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabee3242f8f0b2e4efcb61e8f8307d31300fc14&vid=dabee3242f8f0b2e4efcb61e8f8307d31300fc14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR