6-Hydroxy-1-tetralone ≥98.0%

Supplier: TCI America

H1335-25G H1335-5G
TCH1335-25GEA 477.16 CAD
TCH1335-25G TCH1335-5G
6-Hydroxy-1-tetralone ≥98.0%
6-Hydroxy-1-tetralone

CAS Number: 3470-50-6
MDL Number: MFCD00156670
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 155


Other Product Names / Synonyms: 6-Hydroxy-3,4-dihydronaphthalen-1(2H)-one

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00156670
CAS Number: 3470-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c0956e4854c8e0367f50c4d79a4053b0099d2ca&vid=3c0956e4854c8e0367f50c4d79a4053b0099d2ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR