(R)-(-)-3-Hydroxytetrahydrofuran ≥98.0%

Supplier: TCI America

Synonyms: (R)-Tetrahydrofuran-3-ol

H1332-1G H1332-5G
TCH1332-1GEA 133.83 CAD
TCH1332-1G TCH1332-5G
(R)-(-)-3-Hydroxytetrahydrofuran ≥98.0%
(R)-(-)-3-Hydroxytetrahydrofuran

CAS Number: 86087-24-3
MDL Number: MFCD00067101
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 82
Specific Gravity (20/20): 1.11
Specific rotation [a]20/D: -19 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-Tetrahydrofuran-3-ol

Formula: C₄H₈O₂
Boiling Pt: 80 °C
Density: 1.11 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00067101
CAS Number: 86087-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1896d593030b0f6d429d98cfb0c158ceac7c6a34&vid=1896d593030b0f6d429d98cfb0c158ceac7c6a34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR