(±)-3-Hydroxytetrahydrofuran ≥97.0% (by GC)
Supplier: TCI America
H1331-25G
H1331-5G
TCH1331-25GEA
219.62
CAD
TCH1331-25G
TCH1331-5G
(±)-3-Hydroxytetrahydrofuran ≥97.0% (by GC)
(±)-3-Hydroxytetrahydrofuran
CAS Number: 453-20-3
MDL Number: MFCD00005374
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 81
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: Tetrahydrofuran-3-ol
Formula:
C₄H₈O₂ Boiling Pt: 181 °C Density: 1.11 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00005374 CAS Number: 453-20-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...