Hexyl propionate ≥98.0%

Supplier: TCI America

Synonyms: Propionic Acid Hexyl Ester

H1330-25ML H1330-500ML
TCH1330-25MLEA 66.36 CAD
TCH1330-25ML TCH1330-500ML
Hexyl propionate ≥98.0%
Hexyl propionate

CAS Number: 2445-76-3
MDL Number: MFCD00085197
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 65
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Propionic Acid Hexyl Ester

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 186 °C
Density: 0.87 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00085197
CAS Number: 2445-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f9e0999493e6cc782315121b0d23b40e1224311&vid=7f9e0999493e6cc782315121b0d23b40e1224311&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR