DL-Homophenylalanine ≥97.0%

Supplier: TCI America

Synonyms: 2-Amino-4-phenylbutyric Acid, H-DL-HomoPhe-OH, H-DL-HoPhe-OH

H1329-1G H1329-5G
TCH1329-1GEA 90.06 CAD
TCH1329-1G TCH1329-5G
DL-Homophenylalanine ≥97.0%
DL-Homophenylalanine

CAS Number: 1012-05-1
MDL Number: MFCD00063092
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >97.0% (HPLC,T)
Melting point (°C): 294


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-4-phenylbutyric Acid

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 294 °C
MDL Number: MFCD00063092
CAS Number: 1012-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7478465a05bd23b92219d5f73c426d46423db9bc&vid=7478465a05bd23b92219d5f73c426d46423db9bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR