2-Hydroxy-4-(trifluoromethyl)pyrimidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

H1328-1G H1328-5G
TCH1328-1GEA 167.01 CAD
TCH1328-1G TCH1328-5G
2-Hydroxy-4-(trifluoromethyl)pyrimidine ≥97.0% (by GC, titration analysis)
2-Hydroxy-4-(trifluoromethyl)pyrimidine

CAS Number: 104048-92-2
MDL Number: MFCD00192529
Molecular Formula: C5H3F3N2O
Molecular Weight: 164.09
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 219
Lambda max.: 322 nm (MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)-2-pyrimidinol

Formula: C₅H₃F₃N₂O
Melting Pt: 219 °C
MDL Number: MFCD00192529
CAS Number: 104048-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a33bd1fa4afcfb59bf110e4704333822c22a259&vid=56f39b7663d61a5812035aab572fa206b5f55340&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR