4-Hydroxy-2,6-dimethoxybenzaldehyde ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde

H1326-1G H1326-5G
TCH1326-1GEA 279.66 CAD
TCH1326-1G TCH1326-5G
4-Hydroxy-2,6-dimethoxybenzaldehyde ≥97.0% (by GC, titration analysis)
4-Hydroxy-2,6-dimethoxybenzaldehyde

CAS Number: 22080-96-2
MDL Number: MFCD01407645
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 220

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Melting Pt: 220 °C
MDL Number: MFCD01407645
CAS Number: 22080-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bed319c4a21e1a9df0d3d42412f0e8d80f6cb5a&vid=3bed319c4a21e1a9df0d3d42412f0e8d80f6cb5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR