3-Hydroxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1324-1G H1324-5G
TCH1324-1GEA 93.22 CAD
TCH1324-1G TCH1324-5G
3-Hydroxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Hydroxy-2-nitrobenzoic acid

CAS Number: 602-00-6
MDL Number: MFCD00017003
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₇H₅NO₅
Melting Pt: 182 °C
MDL Number: MFCD00017003
CAS Number: 602-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f6f134049b0cce1a014ba1c520bdac35ac6c760&vid=4f6f134049b0cce1a014ba1c520bdac35ac6c760&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR