5-Hydroxy-2-nitrobenzoic acid ≥98.0%
Supplier: TCI America
H1321-25G
H1321-5G
TCH1321-25GEA
290.88
CAD
TCH1321-25G
TCH1321-5G
5-Hydroxy-2-nitrobenzoic acid ≥98.0%
5-Hydroxy-2-nitrobenzoic acid
CAS Number: 610-37-7
MDL Number: MFCD00017566
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 170
Formula:
C₇H₅NO₅ Melting Pt: 170 °C |
MDL Number:
MFCD00017566 CAS Number: 610-37-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...