4-Hydroxy-3-methylbenzoic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-3-toluic acid, 4-Hydroxy-m-toluic acid

H1319-25G H1319-5G
TCH1319-25GEA 353.92 CAD
TCH1319-25G TCH1319-5G
4-Hydroxy-3-methylbenzoic acid ≥98.0% (by GC)
4-Hydroxy-3-methylbenzoic acid

CAS Number: 499-76-3 MDL Number: MFCD00270105 Molecular Formula: C8H8O3 Molecular Weight: 152.15 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 175


Other Product Names / Synonyms: 4-Hydroxy-m-toluic Acid

Formula: C₈H₈O₃
MW: 152.15
Melting Pt: 175 °C
MDL Number: MFCD00270105
CAS Number: 499-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8def699d96cfefb2d015a97625e4ccf4af8940&vid=cd04e463903ad0a58022c2f5c5d5bce726a0ee4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR