4-(4-Hydroxyphenyl)cyclohexanone ≥95.0%
Supplier: TCI America
H1316-25G
H1316-5G
TCH1316-25GEA
133.83
CAD
TCH1316-25G
TCH1316-5G
4-(4-Hydroxyphenyl)cyclohexanone ≥95.0%
4-(4-Hydroxyphenyl)cyclohexanone
CAS Number: 105640-07-1
MDL Number: MFCD00210693
Molecular Formula: C12H14O2
Molecular Weight: 190.24
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 169
Other Product Names / Synonyms: 4-(4-Oxocyclohexyl)phenol
Formula:
C₁₂H₁₄O₂ Melting Pt: 169 °C |
MDL Number:
MFCD00210693 CAS Number: 105640-07-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...