4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 98,0
Supplier: TCI America
Synonyms:
Zingerone, Vanillylacetone
CAS Number: 122-48-5
MDL Number: MFCD00048232
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 41
Flash Point (°C): 102
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Vanillylacetone, Zingerone
Formula:
C₁₁H₁₄O₃ MW: 194.23 g/mol Boiling Pt: 148 °C Melting Pt: 41 °C Flash Pt: 102 °C |
MDL Number:
MFCD00048232 CAS Number: 122-48-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...