4-Hydroxy-o-toluic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid

H1311-25G H1311-5G
TCH1311-25GEA 266.88 CAD
TCH1311-25G TCH1311-5G
4-Hydroxy-o-toluic acid ≥98.0%
4-Hydroxy-o-toluic acid

CAS Number: 578-39-2
MDL Number: MFCD02182261
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 179


Other Product Names / Synonyms: 4-Hydroxy-o-toluic Acid

Formula: C₈H₈O₃
Melting Pt: 179 °C
MDL Number: MFCD02182261
CAS Number: 578-39-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR