6-Hydroxy-1-indanone ≥98.0%

Supplier: TCI America

Synonyms: 6-Hydroxy-2,3-dihydro-1H-inden-1-one

H1310-5G H1310-1G
TCH1310-5GEA 301.78 CAD
TCH1310-5G TCH1310-1G
6-Hydroxy-1-indanone ≥98.0%
6-Hydroxy-1-indanone

CAS Number: 62803-47-8
MDL Number: MFCD00513530
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 157

Formula: C₉H₈O₂
Melting Pt: 157 °C
MDL Number: MFCD00513530
CAS Number: 62803-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8ffc821b785317b9c8a02016f445edb47c86199&vid=e8ffc821b785317b9c8a02016f445edb47c86199&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR