Honokiol ≥95.0%
Supplier: TCI America
Synonyms:
3',5-Diallyl-2,4'-dihydroxybiphenyl, 3',5-Diallylbiphenyl-2,4'-diol, 5,3'-Diallyl-2,4'-dihydroxybiphenyl
CAS Number: 35354-74-6
MDL Number: MFCD00016674
Molecular Formula: C18H18O2
Molecular Weight: 266.34
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 85
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antitumor Ingredients, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 3',5-Diallylbiphenyl-2,4'-diol, 3',5-Diallyl-2,4'-dihydroxybiphenyl
Formula:
C₁₈H₁₈O₂ Melting Pt: 85 °C |
MDL Number:
MFCD00016674 CAS Number: 35354-74-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...