Hydroxychloroquine sulfate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline sulfate, 2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulphate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulfate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulphate, 2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulfate, 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline Sulphate

H1306-25G H1306-5G
TCH1306-25GEA 327.22 CAD
TCH1306-25G TCH1306-5G
Hydroxychloroquine sulfate ≥98.0% (by HPLC, total nitrogen)
Hydroxychloroquine sulfate

CAS Number: 747-36-4
MDL Number: MFCD00078203
Molecular Formula: C18H26ClN3O
Molecular Weight: 433.95
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Antiprotozoal Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline Sulfate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine Sulfate

Formula: C₁₈H₂₆ClN₃O
MW: 433.95 g/mol
MDL Number: MFCD00078203
CAS Number: 747-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c747e5514af03c8bbacdbca0e98223e81f0b5d81&vid=c747e5514af03c8bbacdbca0e98223e81f0b5d81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR