3-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 850881-09-3
MDL Number: MFCD11045447
Molecular Formula: C16H27BO2S
Molecular Weight: 294.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.99
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hexyl-2-thiopheneboronic Acid Pinacol Ester
Formula:
C₁₆H₂₇BO₂S Density: 0.99 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD11045447 CAS Number: 850881-09-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...