4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0%
Supplier: TCI America
Synonyms:
2-(4-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hexyl-2-thiopheneboronic Acid Pinacol Ester, 2-(4-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS Number: 883742-29-8
Molecular Formula: C16H27BO2S
Molecular Weight: 294.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(4-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hexyl-2-thiopheneboronic Acid Pinacol Ester
Formula:
C₁₆H₂₇BO₂S MW: 294.26 g/mol Density: 0.99 (20 °C) |
MDL Number:
MFCD14708172 CAS Number: 883742-29-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...