2-(Hydroxymethyl)phenylboronic acid cyclic monoester
Supplier: TCI America
H1280-1G
H1280-5G
TCH1280-1GEA
104.28
CAD
TCH1280-1G
TCH1280-5G
2-(Hydroxymethyl)phenylboronic acid cyclic monoester
2-(Hydroxymethyl)phenylboronic acid cyclic monoester
CAS Number: 5735-41-1
MDL Number: MFCD01075677
Molecular Formula: C7H7BO2
Molecular Weight: 133.94
Form: Crystal
Color: White
Melting point (°C): 98
Other Product Names / Synonyms: 1,3-Dihydro-1-hydroxy-2,1-benzoxaborole
Formula:
C₇H₇BO₂ Melting Pt: 98 °C |
MDL Number:
MFCD01075677 CAS Number: 5735-41-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...