3-Methoxysalicyl alcohol ≥98.0% (by GC)

Supplier: TCI America

H1273-1G H1273-5G
TCH1273-1GEA 181.7 CAD
TCH1273-1G TCH1273-5G
3-Methoxysalicyl alcohol ≥98.0% (by GC)
3-Methoxysalicyl alcohol

CAS Number: 4383-05-5
MDL Number: MFCD00008362
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 63


Other Product Names / Synonyms: 2-(Hydroxymethyl)-6-methoxyphenol, 2-Hydroxy-3-(hydroxymethyl)anisole

Formula: C₈H₁₀O₃
Boiling Pt: 162 °C
Melting Pt: 63 °C
MDL Number: MFCD00008362
CAS Number: 4383-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b000f17c4f22fb4f7436bf87f4757dd435e1d9d&vid=3b000f17c4f22fb4f7436bf87f4757dd435e1d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR