4-Hydroxy-2-methoxybenzaldehyde ≥97.0%
Supplier: TCI America
H1191-1G
H1191-25G
H1191-5G
TCH1191-1GEA
82.16
CAD
TCH1191-1G
TCH1191-25G
TCH1191-5G
4-Hydroxy-2-methoxybenzaldehyde ≥97.0%
4-Hydroxy-2-methoxybenzaldehyde
CAS Number: 18278-34-7
MDL Number: MFCD00051964
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 158
Other Product Names / Synonyms: 4-Hydroxy-o-anisaldehyde
Formula:
C₈H₈O₃ MW: 152.15 Boiling Pt: 273 Melting Pt: 158 °C |
MDL Number:
MFCD00051964 CAS Number: 18278-34-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...