5-Hydroxy-3,4-dimethoxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-4,5-dimethoxybenzaldehyde, 3,4-Dimethoxy-5-hydroxybenzaldehyde

H1179-1G H1179-5G
TCH1179-1GEA 82.63 CAD
TCH1179-1G TCH1179-5G
5-Hydroxy-3,4-dimethoxybenzaldehyde ≥98.0%
5-Hydroxy-3,4-dimethoxybenzaldehyde

CAS Number: 29865-90-5
MDL Number: MFCD00010442
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 66


Other Product Names / Synonyms: 5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde

Formula: C₉H₁₀O₄
Boiling Pt: 180 °C
Melting Pt: 66 °C
MDL Number: MFCD00010442
CAS Number: 29865-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e282c74a11f5eed1cee60681de3bf403965624d&vid=8e282c74a11f5eed1cee60681de3bf403965624d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR