2-Hydroxy-1,4-naphthoquinone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Hydroxy-p-naphthoquinone, 2-Hydroxy-1,4-naphoquinone
H0285-25G
H0285-100G
TCH0285-25GEA
66.36
CAD
TCH0285-25G
TCH0285-100G
2-Hydroxy-1,4-naphthoquinone ≥98.0% (by titrimetric analysis)
2-Hydroxy-1,4-naphthoquinone
CAS Number: 83-72-7
MDL Number: MFCD00001678
Molecular Formula: C10H6O3
Molecular Weight: 174.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₁₀H₆O₃ MW: 174.16 g/mol |
MDL Number:
MFCD00001678 CAS Number: 83-72-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...