Guanidine hydroiodide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
G0450-1G
G0450-5G
TCG0450-1GEA
37.92
CAD
TCG0450-1G
TCG0450-5G
Guanidine hydroiodide ≥97.0% (by titrimetric analysis)
Guanidine hydroiodide
CAS Number: 19227-70-4
Molecular Formula: CH5N3
Molecular Weight: 186.98
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 196
Formula:
CH₅N₃ Melting Pt: 196 °C |
CAS Number:
19227-70-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...