Glycidyl lauryl ether ≥98.0%

Supplier: TCI America

Synonyms: Dodecyl Glycidyl Ether

G0448-1G G0448-5G
TCG0448-1GEA 115.34 CAD
TCG0448-1G TCG0448-5G
Glycidyl lauryl ether ≥98.0%
Glycidyl lauryl ether

CAS Number: 2461-18-9
MDL Number: MFCD00022344
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 324
Specific Gravity (20/20): 0.88


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Dodecyl Glycidyl Ether

Formula: C₁₅H₃₀O₂
MW: 242.40 g/mol
Boiling Pt: 324 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00022344
CAS Number: 2461-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0c7e3930607a6f17e39866c11af9fc419576f16&vid=f0c7e3930607a6f17e39866c11af9fc419576f16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR