Glycine sulfate ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: Glycine sulphate, Glycine Sulfate, H-Gly-OH.1/3H2SO4

G0424-25G G0424-5G
TCG0424-25GEA 619.36 CAD
TCG0424-25G TCG0424-5G
Glycine sulfate ≥98.0% (by total nitrogen and titration analysis)
Glycine sulfate

CAS Number: 513-29-1
MDL Number: MFCD00036426
Molecular Formula: C2H5NO2
Molecular Weight: 107.76
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Gly-OH.1/3H2SO4

Formula: C₂H₅NO₂
MW: 107.76 g/mol
MDL Number: MFCD00036426
CAS Number: 513-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1d4ac0a4d2257d1d980abd99babd19a263db365&vid=f1d4ac0a4d2257d1d980abd99babd19a263db365&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR