1-(2-Methoxyphenoxy)-2,3-epoxypropane ≥98.0% (by GC)

Supplier: TCI America

G0416-25G G0416-5G
TCG0416-25GEA 311.26 CAD
TCG0416-25G TCG0416-5G
1-(2-Methoxyphenoxy)-2,3-epoxypropane ≥98.0% (by GC)
1-(2-Methoxyphenoxy)-2,3-epoxypropane

CAS Number: 2210-74-4
MDL Number: MFCD00190158
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 39


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Glycidyl Guaiacol Ether, 1,2-Epoxy-3-(2-methoxyphenoxy)propane

Formula: C₁₀H₁₂O₃
Boiling Pt: 120 °C
Melting Pt: 39 °C
MDL Number: MFCD00190158
CAS Number: 2210-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=690c900234e4289e686d7e4ef64e8b62b9378324&vid=690c900234e4289e686d7e4ef64e8b62b9378324&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR