(S)-α-(2-Oxiranyl)anisole ≥98.0%

Supplier: TCI America

Synonyms: (S)-Glycidyl phenyl ether

G0410-1G G0410-5G
TCG0410-1GEA 60.04 CAD
TCG0410-1G TCG0410-5G
(S)-α-(2-Oxiranyl)anisole ≥98.0%
(S)-α-(2-Oxiranyl)anisole

CAS Number: 71031-03-3
MDL Number: MFCD06659026
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.11
Specific rotation [a]20/D: 14 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-(Phenoxymethyl)oxirane, (S)-1,2-Epoxy-3-phenoxypropane

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
Density: 1.11 (20 °C)
MDL Number: MFCD06659026
CAS Number: 71031-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f2587bfbfd9047fc6fd6ffead98b72bc0d5bf4f&vid=9f2587bfbfd9047fc6fd6ffead98b72bc0d5bf4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR