2-O-ɑ-D-glucopyranosyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

G0394-1G G0394-5G
TCG0394-1GEA 50.56 CAD
TCG0394-1G TCG0394-5G
2-O-ɑ-D-glucopyranosyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
2-O-alpha-D-glucopyranosyl-L-ascorbic acid

CAS Number: 129499-78-1
MDL Number: MFCD00925623
Molecular Formula: C12H18O11
Molecular Weight: 338.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160
Specific rotation [a]20/D: 193 deg (C=1, H2O)
Lambda max.: 260 nm (H2O)


Product Categories: Bioscience, Glycosides, Disaccharides, Vitamin Derivatives
Other Product Names / Synonyms: AA-2G

Formula: C₁₂H₁₈O₁₁
Melting Pt: 160 °C
MDL Number: MFCD23701380
CAS Number: 129499-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56c5c43b9180330344db82a4c0e84cefb4b9640&vid=e3280657bb2f10bbdf5e3795a05c30881a319ba0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR