(+)-Griseofulvin ≥97.0%

Supplier: TCI America

Synonyms: Griseofulvin

G0384-25G G0384-5G
TCG0384-25GEA 152.55 CAD
TCG0384-25G TCG0384-5G
(+)-Griseofulvin ≥97.0%
(+)-Griseofulvin

CAS Number: 126-07-8
MDL Number: MFCD00082343
Molecular Formula: C17H17ClO6
Molecular Weight: 352.77
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 220
Specific rotation [a]20/D: 360 deg (C=1, DMF)
Lambda max.: 321 nm (CHCl3)Storage Temperature: 0-10°C

Formula: C₁₇H₁₇ClO₆
MW: 352.77 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00082343
CAS Number: 126-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6693a865b4d11ce03563974fd2ab607afe46c043&vid=6693a865b4d11ce03563974fd2ab607afe46c043&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR