Glibenclamide ≥98.5% (by HPLC, titration analysis)

Supplier: TCI America

G0382-25G G0382-5G
TCG0382-25GEA 135.88 CAD
TCG0382-25G TCG0382-5G
Glibenclamide ≥98.5% (by HPLC, titration analysis)
Glibenclamide

CAS Number: 10238-21-8
MDL Number: MFCD00056625
Molecular Formula: C23H28ClN3O5S
Molecular Weight: 494.00
Purity/Analysis Method: >98.5% (HPLC,T)
Form: Crystal
Melting point (°C): 174
Storage Temperature: 0-10°C


Product Categories: Bioscience, Obesity/Diabete Research Reagents, Antidiabetic Ingredients
Other Product Names / Synonyms: 5-Chloro-N-[4-(3-cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide

Formula: C₂₃H₂₈ClN₃O₅S
Melting Pt: 174 °C
MDL Number: MFCD00056625
CAS Number: 10238-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c17e5b62112948046c51df554fb4d5cee904f382&vid=c17e5b62112948046c51df554fb4d5cee904f382&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR