β-D-Glucopyranose-1-phosphate disodium salt ≥98.0%

Supplier: TCI America

G0339-100MG G0339-20MG
TCG0339-100MGEA 3152.1 CAD
TCG0339-100MG TCG0339-20MG
β-D-Glucopyranose-1-phosphate disodium salt ≥98.0%
beta-D-Glucopyranose-1-phosphate disodium salt

CAS Number: 83833-15-2
Molecular Formula: C6H13O9P
Molecular Weight: 304.10
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C

Formula: C₆H₁₃O₉P
MW: 304.10 g/mol
CAS Number: 83833-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61af2022bfb5089ac2ae86a5562b651843737fa2&vid=61af2022bfb5089ac2ae86a5562b651843737fa2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR