Gal[2346Ac]β(1-3)GalN3[46Bzd]-β-MP ≥95.0% (by HPLC)

Supplier: TCI America

G0330-1G G0330-5G
TCG0330-1GEA 619.83 CAD
TCG0330-1G TCG0330-5G
Gal[2346Ac]β(1-3)GalN3[46Bzd]-β-MP ≥95.0% (by HPLC)
Gal[2346Ac]beta(1-3)GalN3[46Bzd]-beta-MP

Molecular Weight: 729.69
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 193
Specific rotation [a]20/D: 11 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Molecular Formula: C34H39N3O15


Other Product Names / Synonyms: 4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranosyl)-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-galactopyranoside

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f54cf6df7726f9ac8fb60638efa823a5f07ab6&vid=458f9287c113145b05f838843d2b25aaf4ab0f1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR