Gal[2346Ac]β(1-3)GlcNPhth[46Bzd]-β-MP ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-4,6-O-benzylidene-2-deoxy-2-phthalimido-β-D-glucopyranoside
Molecular Weight: 833.80
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 12 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Molecular Formula: C42H43NO17
Other Product Names / Synonyms: 4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranosyl)-4,6-O-benzylidene-2-deoxy-2-phthalimido-beta-D-glucopyranoside
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...