Gal[2346Ac]β1-3)GlcN3[46Bzd]-β-MP ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-2-azido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranoside

G0309-1G G0309-5G
TCG0309-1GEA 352.02 CAD
TCG0309-1G TCG0309-5G
Gal[2346Ac]β1-3)GlcN3[46Bzd]-β-MP ≥98.0% (by HPLC)
Gal[2346Ac]beta(1-3)GlcN3[46Bzd]-beta-MP

Molecular Weight: 729.69
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 172
Specific rotation [a]20/D: -13 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Molecular Formula: C34H39N3O15


Other Product Names / Synonyms: 4-Methoxyphenyl 3-O-(2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranosyl)-2-azido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranoside

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c281dda80ebc4c8f55f9739bbd457ad6a0c83a9d&vid=5940be877bf4df80b8ee1d5c9f358d2def4ce1f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR