GlcNPhth[346Ac]-β-(1-3)Gal[246Bn]-β-MP ≥98.0%
Supplier: TCI America
Synonyms:
4-Methoxyphenyl 3-O-(3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranosyl)-2,4,6-tri-O-benzyl-β-D-galactopyranoside
Molecular Weight: 974.03
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 175
Specific rotation [a]20/D: -8 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Molecular Formula: C54H55NO16
Other Product Names / Synonyms: 4-Methoxyphenyl 3-O-(3,4,6-Tri-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranosyl)-2,4,6-tri-O-benzyl-beta-D-galactopyranoside
Formula:
C₅₄H₅₅NO₁₆ Melting Pt: 175 °C |
CAS Number:
1820575-44-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...