Disodium-α-glycerophosphate hydrate ≥85.0% (by titrimetric analysis)
Supplier: TCI America
G0096-25G
G0096-5G
TCG0096-25GEA
570.38
CAD
TCG0096-25G
TCG0096-5G
Disodium-α-glycerophosphate hydrate ≥85.0% (by titrimetric analysis)
Disodium α-glycerophosphate
CAS Number: 1555-56-2
MDL Number: MFCD00035539
Molecular Formula: C3H9O6P
Molecular Weight: 216.04
Purity/Analysis Method:
Other Product Names / Synonyms: alpha-Glycerophosphoric Acid Disodium Salt
Formula:
C₃H₇Na₂O₆P·xH₂O |
MDL Number:
MFCD00035539 CAS Number: 1555-56-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...