5-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0976-1G
F0976-5G
TCF0976-1GEA
74.26
CAD
TCF0976-1G
TCF0976-5G
5-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-methylphenylboronic acid
CAS Number: 163517-62-2
MDL Number: MFCD03095047
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Form: Crystal
Color: White
Melting point (°C): 148
Other Product Names / Synonyms: 5-Fluoro-2-methylbenzeneboronic Acid
Formula:
C₇H₈BFO₂ Melting Pt: 148 °C |
MDL Number:
MFCD03095047 CAS Number: 163517-62-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...