N-(2-Fluorophenyl)-3-oxobutyramide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

F0963-1G F0963-200MG
TCF0963-1GEA 413.96 CAD
TCF0963-1G TCF0963-200MG
N-(2-Fluorophenyl)-3-oxobutyramide ≥98.0% (by total nitrogen basis)
N-(2-Fluorophenyl)-3-oxobutyramide

CAS Number: 5279-85-6
MDL Number: MFCD00661533
Molecular Formula: C10H10FNO2
Molecular Weight: 195.19
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 74


Product Categories: Synthetic Organic Chemistry, Diketone Compounds (Multi-component Reaction), ƒÀ-Keto Esters (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 2'-Fluoroacetoacetanilide

Formula: C₁₀H₁₀FNO₂
Melting Pt: 74 °C
MDL Number: MFCD00661533
CAS Number: 5279-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b774d5371920f2ffbe0bc2d029ffb3d1947ade99&vid=b774d5371920f2ffbe0bc2d029ffb3d1947ade99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR